Assay Details
Gene Symbol: FBXL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040015.1 | XP_017895504.1 | 7 - 8 | 2252 | 52 | |
XM_018040016.1 | XP_017895505.1 | 7 - 8 | 2216 | 52 | ||
XM_018040017.1 | XP_017895506.1 | 7 - 8 | 2123 | 52 | ||
XM_018040018.1 | XP_017895507.1 | 5 - 6 | 883 | 52 | ||
XM_018040019.1 | XP_017895508.1 | 5 - 6 | 754 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
