Assay Details
| Gene Symbol: HSD3B7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974644.2 | XP_013830098.2 | 6 - 7 | 1031 | 58 | |
| XM_018040026.1 | XP_017895515.1 | 6 - 7 | 979 | 58 | ||
| GenBank mRNA | KA348258.1 | - | 6 - 7 | 894 | 58 | |
More Information
Set Membership: |
|

HSD3B7
Genomic Map