Assay Details
Gene Symbol: ZNF436 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676867.3 | XP_005676924.1 | 3 - 4 | 679 | 72 | |
XM_005676869.3 | XP_005676926.1 | 2 - 3 | 217 | 72 | ||
XM_005676870.3 | XP_005676927.1 | 2 - 3 | 586 | 72 | ||
XM_018055558.1 | XP_017911047.1 | 2 - 3 | 531 | 72 | ||
XM_018055572.1 | XP_017911061.1 | 2 - 3 | 611 | 72 | ||
GenBank mRNA | JO513883.2 | - | 1 - 2 | 582 | 72 | |
JT749039.1 | - | 3 - 4 | 440 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
