Assay Details
| Gene Symbol: BET1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678935.3 | XP_005678992.1 | 3 - 4 | 373 | 100 | |
| GenBank mRNA | JO450264.2 | - | 1 - 2 | 322 | 100 | |
More Information
Set Membership: |
|

BET1
Genomic Map