Assay Details
Gene Symbol: STIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049352.1 | XP_017904841.1 | 7 - 8 | 1615 | 83 | |
XM_018049353.1 | XP_017904842.1 | 7 - 8 | 1612 | 83 | ||
GenBank mRNA | JT663103.1 | - | 1811 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
