Assay Details
Gene Symbol: GEMIN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686517.3 | XP_005686574.1 | 2 - 3 | 370 | 92 | |
XM_013967565.2 | XP_013823019.1 | 2 - 3 | 200 | 92 | ||
GenBank mRNA | JO336222.2 | - | 1 - 2 | 349 | 92 | |
JO451458.2 | - | 1 - 2 | 458 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
