Assay Details
| Gene Symbol: PTRHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686944.3 | XP_005687001.1 | 1 - 2 | 278 | 83 | |
| GenBank mRNA | JO383583.2 | - | 1 - 2 | 257 | 83 | |
| JT680477.1 | - | 3 - 4 | 620 | 83 | ||
More Information
Set Membership: |
|

PTRHD1
Genomic Map