Assay Details
Gene Symbol: SYT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059486.1 | XP_017914975.1 | 2 - 3 | 719 | 70 | |
XM_018059487.1 | XP_017914976.1 | 2 - 3 | 719 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
