Assay Details
| Gene Symbol: ALKBH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691590.3 | XP_005691647.3 | 3 - 4 | 990 | 74 | |
| XM_013970768.2 | XP_013826222.2 | 3 - 4 | 723 | 74 | ||
| XM_013970769.2 | XP_013826223.2 | 3 - 4 | 711 | 74 | ||
| XM_013970770.2 | XP_013826224.2 | 3 - 4 | 692 | 74 | ||
| GenBank mRNA | JT710237.1 | - | 3 - 4 | 208 | 74 | |
More Information
Set Membership: |
|

ALKBH2
Genomic Map