Assay Details
Gene Symbol: APOBEC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696288.3 | XP_005696345.2 | 1 - 2 | 287 | 90 | |
GenBank mRNA | JO366318.2 | - | 1 - 2 | 327 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
