Assay Details
| Gene Symbol: LOC102183469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696368.3 | XP_005696425.2 | 7 - 8 | 1211 | 103 | |
| XM_013973925.2 | XP_013829379.2 | 6 - 7 | 727 | 103 | ||
| GenBank mRNA | JO434721.2 | - | 4 - 5 | 348 | 103 | |
More Information
Set Membership: |
|

LOC102183469
Genomic Map