Assay Details
| Gene Symbol: KCTD14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699495.3 | XP_005699552.1 | 1 - 2 | 113 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCTD14| Gene Symbol: KCTD14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699495.3 | XP_005699552.1 | 1 - 2 | 113 | 81 | |
Set Membership: |
|