Assay Details
| Gene Symbol: LOC102183271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO631943.2 | - | 1606 | 74 | ||
| Gene Symbol: APBA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052054.1 | XP_017907543.1 | 3 - 4 | 1558 | 74 | |
| XM_018052056.1 | XP_017907545.1 | 3 - 4 | 1453 | 74 | ||
More Information
Set Membership: |
|

LOC102183271
Genomic Map