Assay Details
Gene Symbol: LRFN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063313.1 | XP_017918802.1 | 3 - 4 | 2579 | 61 | |
XM_018063314.1 | XP_017918803.1 | 3 - 4 | 2576 | 61 | ||
XM_018063315.1 | XP_017918804.1 | 2 - 3 | 2476 | 61 | ||
XM_018063316.1 | XP_017918805.1 | 2 - 3 | 2473 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
