Assay Details
Gene Symbol: MAJIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013975960.2 | XP_013831414.2 | 5 - 6 | 375 | 55 | |
XM_018043595.1 | XP_017899084.1 | 8 - 9 | 609 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
