Assay Details
Gene Symbol: MAP3K7CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674712.3 | XP_005674769.1 | 5 - 6 | 632 | 62 | |
XM_013964618.2 | XP_013820072.1 | 4 - 5 | 576 | 62 | ||
XM_013964647.2 | XP_013820101.1 | 4 - 5 | 765 | 62 | ||
GenBank mRNA | JT737996.1 | - | 1 - 2 | 424 | 62 | |
KA310315.1 | - | 4 - 5 | 565 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
