Assay Details
| Gene Symbol: PYCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688914.3 | XP_005688971.2 | 4 - 5 | 619 | 69 | |
| GenBank mRNA | JO434108.2 | - | 4 - 5 | 584 | 69 | |
| KA331420.1 | - | 2 - 3 | 826 | 69 | ||
More Information
Set Membership: |
|

PYCR3
Genomic Map