Assay Details
| Gene Symbol: NOCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061536.1 | XP_017917025.1 | 2 - 3 | 859 | 72 | |
| XM_018061537.1 | XP_017917026.1 | 3 - 4 | 708 | 72 | ||
More Information
Set Membership: |
|

NOCT
Genomic Map