Assay Details
Gene Symbol: RPP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695789.3 | XP_005695846.2 | 2 - 3 | 406 | 64 | |
XM_005695790.3 | XP_005695847.2 | 2 - 3 | 262 | 64 | ||
XM_005695791.3 | XP_005695848.2 | 2 - 3 | 242 | 64 | ||
XM_018066773.1 | XP_017922262.1 | 2 - 3 | 430 | 64 | ||
GenBank mRNA | JO599586.2 | - | 2 - 3 | 379 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
