Assay Details
| Gene Symbol: AP3S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053721.1 | XP_017909210.1 | 4 - 5 | 603 | 88 | |
| XM_018053722.1 | XP_017909211.1 | 4 - 5 | 603 | 88 | ||
| GenBank mRNA | KA319416.1 | - | 2 - 3 | 811 | 88 | |
More Information
Set Membership: |
|

AP3S1
Genomic Map