Assay Details
| Gene Symbol: GTF3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968228.2 | XP_013823682.2 | 4 - 5 | 694 | 65 | |
| GenBank mRNA | JO612057.2 | - | 4 - 5 | 586 | 65 | |
More Information
Set Membership: |
|

GTF3A
Genomic Map