Assay Details
| Gene Symbol: TNFRSF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059943.1 | XP_017915432.1 | 12 - 13 | 1386 | 60 | |
| GenBank mRNA | JO614913.2 | - | 1 - 2 | 1085 | 60 | |
| JO614914.2 | - | 1 - 2 | 1085 | 60 | ||
More Information
Set Membership: |
|

TNFRSF8
Genomic Map