Assay Details
Gene Symbol: ITM2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058402.1 | XP_017913891.1 | 3 - 4 | 596 | 66 | |
GenBank mRNA | JT748783.1 | - | 3 - 4 | 526 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
