Assay Details
| Gene Symbol: ACOT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043922.1 | XP_017899411.1 | 7 - 8 | 740 | 73 | |
| GenBank mRNA | JO384643.2 | - | 8 - 9 | 974 | 73 | |
More Information
Set Membership: |
|

ACOT9
Genomic Map