Assay Details
Gene Symbol: SUMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052124.1 | XP_017907613.1 | 2 - 3 | 354 | 71 | |
GenBank mRNA | JO611640.2 | - | 2 - 3 | 310 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
