Assay Details
Gene Symbol: LOC102191423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013965702.2 | XP_013821156.1 | 3 - 4 | 261 | 67 | |
XM_013965703.2 | XP_013821157.1 | 3 - 4 | 366 | 67 | ||
XM_013965704.2 | XP_013821158.1 | 5 - 6 | 563 | 67 | ||
XM_013965705.2 | XP_013821159.1 | 4 - 5 | 483 | 67 | ||
XM_018050132.1 | XP_017905621.1 | 3 - 4 | 366 | 67 | ||
XM_018050133.1 | XP_017905622.1 | 3 - 4 | 366 | 67 | ||
XM_018050134.1 | XP_017905623.1 | 3 - 4 | 366 | 67 | ||
XM_018050135.1 | XP_017905624.1 | 3 - 4 | 366 | 67 | ||
GenBank mRNA | JT714845.1 | - | 5 - 6 | 493 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
