Assay Details
Gene Symbol: LOC102191423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013965702.2 | XP_013821156.1 | 5 - 6 | 445 | 81 | |
XM_013965703.2 | XP_013821157.1 | 5 - 6 | 550 | 81 | ||
XM_013965704.2 | XP_013821158.1 | 7 - 8 | 747 | 81 | ||
XM_013965705.2 | XP_013821159.1 | 6 - 7 | 667 | 81 | ||
XM_018050132.1 | XP_017905621.1 | 5 - 6 | 550 | 81 | ||
XM_018050133.1 | XP_017905622.1 | 5 - 6 | 550 | 81 | ||
XM_018050134.1 | XP_017905623.1 | 5 - 6 | 550 | 81 | ||
XM_018050135.1 | XP_017905624.1 | 5 - 6 | 550 | 81 | ||
GenBank mRNA | JT714845.1 | - | 7 - 8 | 666 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
