Assay Details
Gene Symbol: LOC102191423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013965702.2 | XP_013821156.1 | 6 - 7 | 542 | 93 | |
XM_013965703.2 | XP_013821157.1 | 6 - 7 | 647 | 93 | ||
XM_013965704.2 | XP_013821158.1 | 8 - 9 | 844 | 93 | ||
XM_013965705.2 | XP_013821159.1 | 7 - 8 | 764 | 93 | ||
XM_018050132.1 | XP_017905621.1 | 6 - 7 | 647 | 93 | ||
XM_018050133.1 | XP_017905622.1 | 6 - 7 | 647 | 93 | ||
XM_018050134.1 | XP_017905623.1 | 6 - 7 | 647 | 93 | ||
XM_018050135.1 | XP_017905624.1 | 6 - 7 | 647 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
