Assay Details
| Gene Symbol: CACUL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013975275.2 | XP_013830729.2 | 5 - 6 | 1105 | 71 | |
| GenBank mRNA | JO588405.2 | - | 5 - 6 | 514 | 71 | |
More Information
Set Membership: |
|

CACUL1
Genomic Map