Assay Details
| Gene Symbol: LTO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043273.1 | XP_017898762.1 | 4 - 5 | 470 | 120 | |
| XM_018043274.1 | XP_017898763.1 | 5 - 6 | 637 | 120 | ||
| XM_018043275.1 | XP_017898764.1 | 4 - 5 | 492 | 120 | ||
| GenBank mRNA | JT668934.1 | - | 2 - 3 | 304 | 120 | |
More Information
Set Membership: |
|

LTO1
Genomic Map