Assay Details
| Gene Symbol: SLC23A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676557.3 | XP_005676614.1 | 11 - 12 | 1775 | 55 | |
| XM_005676558.3 | XP_005676615.1 | 10 - 11 | 1421 | 55 | ||
| GenBank mRNA | JO436542.2 | - | 1 - 2 | 809 | 55 | |
More Information
Set Membership: |
|

SLC23A3
Genomic Map