Assay Details
Gene Symbol: C3H1orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677439.3 | XP_005677496.2 | 4 - 5 | 718 | 62 | |
XM_018046187.1 | XP_017901676.1 | 3 - 4 | 616 | 62 | ||
XM_018046188.1 | XP_017901677.1 | 4 - 5 | 718 | 62 | ||
XM_018046189.1 | XP_017901678.1 | 3 - 4 | 616 | 62 | ||
GenBank mRNA | JO373438.2 | - | 1158 | 62 | ||
JO499345.2 | - | 3 - 4 | 1167 | 62 |
Gene Symbol: LOC102177858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040521.1 | XP_017896010.1 | 1 | 517 | 62 | |
XM_018040522.1 | XP_017896011.1 | 2 | 415 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
