Assay Details
Gene Symbol: SHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682562.3 | XP_005682619.2 | 3 - 4 | 1062 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682562.3 | XP_005682619.2 | 3 - 4 | 1062 | 67 |
Set Membership: |
![]() ![]() ![]() ![]() |