Assay Details
| Gene Symbol: UCMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687951.3 | XP_005688008.1 | 2 - 3 | 177 | 63 | |
| GenBank mRNA | JO416756.2 | - | 2 - 3 | 141 | 63 | |
More Information
Set Membership: |
|

UCMA
Genomic Map