Assay Details
| Gene Symbol: APOA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689482.3 | XP_005689539.3 | 2 - 3 | 441 | 95 | |
| GenBank mRNA | JO420626.2 | - | 1 - 2 | 1135 | 95 | |
More Information
Set Membership: |
|

APOA4
Genomic Map