Assay Details
Gene Symbol: CIB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695085.3 | XP_005695142.1 | 4 - 5 | 465 | 71 | |
XM_005695088.3 | XP_005695145.1 | 3 - 4 | 421 | 71 | ||
XM_018066400.1 | XP_017921889.1 | 5 - 6 | 534 | 71 | ||
XM_018066401.1 | XP_017921890.1 | 5 - 6 | 435 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
