Assay Details
Gene Symbol: COQ7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697529.3 | XP_005697586.1 | 4 - 5 | 569 | 72 | |
XM_005697530.3 | XP_005697587.1 | 4 - 5 | 494 | 72 | ||
GenBank mRNA | JO373199.2 | - | 4 - 5 | 568 | 72 | |
JO445767.2 | - | 4 - 5 | 558 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
