Assay Details
| Gene Symbol: CCSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013964534.2 | XP_013819988.1 | 8 - 9 | 2419 | 77 | |
| XM_018049307.1 | XP_017904796.1 | 8 - 9 | 2419 | 77 | ||
| XM_018049308.1 | XP_017904797.1 | 8 - 9 | 2307 | 77 | ||
More Information
Set Membership: |
|

CCSER1
Genomic Map