Assay Details
| Gene Symbol: PRKACA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051193.1 | XP_017906682.1 | 6 - 7 | 547 | 54 | |
| GenBank mRNA | JO556624.2 | - | 4 - 5 | 524 | 54 | |
More Information
Set Membership: |
|

PRKACA
Genomic Map