Assay Details
| Gene Symbol: LOC102186858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051016.1 | XP_017906505.1 | 9 - 10 | 993 | 87 | |
| GenBank mRNA | JO503309.2 | - | 2 - 3 | 246 | 87 | |
More Information
Set Membership: |
|

LOC102186858
Genomic Map