Assay Details
Gene Symbol: CNNM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013967424.2 | XP_013822878.2 | 6 - 7 | 2115 | 61 | |
XM_018054868.1 | XP_017910357.1 | 6 - 7 | 2117 | 61 | ||
GenBank mRNA | KA305288.1 | - | 6 - 7 | 1266 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
