Assay Details
| Gene Symbol: FHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061609.1 | XP_017917098.1 | 10 - 11 | 2217 | 67 | |
| GenBank mRNA | KA315583.1 | - | 9 - 10 | 938 | 67 | |
More Information
Set Membership: |
|

FHDC1
Genomic Map