Assay Details
Gene Symbol: GLOD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013971906.2 | XP_013827360.1 | 2 - 3 | 214 | 64 | |
XM_018064368.1 | XP_017919857.1 | 2 - 3 | 232 | 64 | ||
XM_018064369.1 | XP_017919858.1 | 3 - 4 | 321 | 64 | ||
GenBank mRNA | JO434044.2 | - | 7 - 8 | 1351 | 64 | |
JO622378.2 | - | 2 - 3 | 229 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018064369.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
