Assay Details
| Gene Symbol: SYCP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038503.1 | XP_017893992.1 | 20 - 21 | 1734 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SYCP2L| Gene Symbol: SYCP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038503.1 | XP_017893992.1 | 20 - 21 | 1734 | 58 | |
Set Membership: |
|