Assay Details
| Gene Symbol: PCNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013975550.2 | XP_013831004.2 | 4 - 5 | 807 | 100 | |
| GenBank mRNA | JO324224.2 | - | 1 - 2 | 252 | 100 | |
| JO324225.2 | - | 1 - 2 | 252 | 100 | ||
| JO363573.2 | - | 4 - 5 | 462 | 100 | ||
| JO394550.2 | - | 1 - 2 | 424 | 100 | ||
| JO451355.2 | - | 1 - 2 | 244 | 100 | ||
| JO498946.2 | - | 673 | 100 | |||
| JO508115.2 | - | 2 - 3 | 279 | 100 | ||
| JO556981.2 | - | 4 - 5 | 492 | 100 | ||
| JO556982.2 | - | 4 - 5 | 489 | 100 | ||
| JO610616.2 | - | 1 - 2 | 243 | 100 | ||
| KA310411.1 | - | 1 - 2 | 947 | 100 | ||
More Information
Set Membership: |
|

PCNP
Genomic Map