Assay Details
Gene Symbol: TMEM207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675121.3 | XP_005675178.2 | 1 - 2 | 191 | 74 | |
GenBank mRNA | JO455030.2 | - | 4 - 5 | 361 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
