Assay Details
| Gene Symbol: CIART | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677679.3 | XP_005677736.1 | 3 - 4 | 1147 | 77 | |
| XM_005677680.3 | XP_005677737.1 | 4 - 5 | 680 | 77 | ||
| XM_018046030.1 | XP_017901519.1 | 4 - 5 | 360 | 77 | ||
More Information
Set Membership: |
|

CIART
Genomic Map