Assay Details
Gene Symbol: CCDC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678558.3 | XP_005678615.1 | 4 - 5 | 834 | 57 | |
XM_018043219.1 | XP_017898708.1 | 4 - 5 | 831 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
