Assay Details
| Gene Symbol: TSPAN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681395.3 | XP_005681452.1 | 2 - 3 | 562 | 60 | |
| GenBank mRNA | JO637459.2 | - | 2 - 3 | 527 | 60 | |
| JT663006.1 | - | 2 - 3 | 478 | 60 | ||
More Information
Set Membership: |
|

TSPAN5
Genomic Map