Assay Details
Gene Symbol: CD68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693517.3 | XP_005693574.1 | 3 - 4 | 1117 | 70 | |
GenBank mRNA | JO579519.1 | - | 2 - 3 | 1382 | 70 | |
JO579520.1 | - | 3 - 4 | 1843 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
